Learn More
Description
Calmodulin (CaM) is a calcium-binding protein found in all eukaryotic cells. It belongs to a structurally homologous family of proteins which includes troponin C, parvalbumin, and S-100. CaM is highly conserved among species, and all vertebrate CaMs have identical structures. Although CaM exists in abundance in many cells, it can be regulated by hormones and neurotransmitters in a variety of tissues and systems.
Host Species: Mouse
Clone: 1F11 + 2D1 + 6D4
Isotype: IgG1 + IgG1 + IgG1
Cross Reactivity: Human, Cow, Rat, Chicken, and Dictyostelium discoideum. Others not tested.
Epitope: Not determined
Immunogen: Purified calmodulin from Dictyostelium discoideum strain.
Molecular Weight: 17kDa
Positive Control: Dictyostelium lysate on IMR5 cells.
Cellular Localization: Cytoplasmic
Ab-4 is a cocktail of Ab-1, Ab-2, and Ab-3.
Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
| Antigen | Calmodulin Ab-4 |
| Classification | Monoclonal |
| Clone | 1F11 + 2D1 + 6D4 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Form | Purified |
| Isotype | IgG1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.