Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calnexin Mouse, Unlabeled, Clone: 37, BD

Catalog No. BDB610523 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610523 50 μg
BDB610524 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610523 Supplier BD Biosciences Supplier No. 610523
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calnexin is a 90kDa integral membrane protein located primarily in the ER. It contains a long N-terminal calcium-binding domain that extends into the lumen of the ER and a short, acidic cytosolic domain. Calnexin associates with several cell surface proteins as they pass through the ER. Since it also forms complexes with other resident ER proteins involved in the Ca2+-dependent retention of proteins, calnexin may act as a chaperone. The amino acid sequence of calnexin is highly conserved among species and is similar in sequence to calreticulin, another Ca2+-binding protein found in the ER. Neither calnexin nor calreticulin contain the calcium-binding motif, known as the “E-F hand” found in the calmodulin family of proteins.

Host Species: Mouse
Clone: 37
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human Calnein aa. 116-301

Immunofluorescence, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen Calnexin
Applications Western Blot
Classification Monoclonal
Clone 37
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.