Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpain Mouse anti-Bovine, Human, Clone: P1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB3083
Description
This Anti-Calpain Antibody, small subunit of μ-or m-Calpains (Calpain I or II), clone P1 is validated for use in ELISA, western blot, immunocytochemistry, immunohistochemistry for the detection of Calpain.
Specifications
Calpain | |
Monoclonal | |
Unconjugated | |
NM_001003962.1; NM_001749.2 | |
Mouse | |
100 μL | |
Bovine, Human | |
Ascites |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
P1 | |
Liquid. Contains no preservative. | |
CAPNS1, CDPS, 30K, CANPS, CAPNS, CANP, CSS1, CAPN4, CALPAIN4 | |
Bovine myocardial m-calpain. | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction