Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain I Mouse anti-Human, Hamster, Mouse, Rat, Clone: P9, MilliporeSigma™

Catalog No. MAB3104MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3104MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3104MI Supplier MilliporeSigma Supplier No. MAB3104

Mouse Monoclonal Antibody

Anti-Calpain I Antibody, large subunit, clone P9 is an antibody against Calpain I for use in ELISA, immunoprecipitation, western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain I
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone P9
Conjugate Unconjugated
Formulation Liquid
Gene Accession No. NM_005186.2
Gene Symbols CAPN1, CANPL1, muCANP, CANP, muCL, Micromolar-calpain, EC 3.4.22.52
Host Species Mouse
Immunogen Calpain-I large subunit from human erythrocyte calpain.
Quantity 100 μL
Primary or Secondary Primary
Target Species Hamster, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.