Learn More
Calpain II Rabbit anti-Rat, Mouse, Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Calpain II |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS pH 7.4 containing 0.02% sodium azide, 1% BSA and 50% glycerol as a stabilizer. |
| Gene Symbols | CAPN2; millimolar-calpain; M-calpain; Millimolar-calpain; mCANP; FLJ39928; CANP2; CANPml; CANPL2; EC 3.4.22.53 |
| Host Species | Rabbit |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.