Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CALR, Clone: 1G11-1A9, Abnova™

Catalog No. 89016412 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-412 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-412 Supplier Abnova Corporation Supplier No. H00000811M01
Add to Cart
Add to Cart

Mouse Monoclonal

Host Species: Mouse
Clone: 1G11-1A9
Isotype: IgG1, κ
Cross Reactivity: Human
Immunogen: CALR (AAH02500, 1 a.a. ∽339 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26kDa

ELISA, Immunohistochemistry, Sandwich ELISA, Western Blotting

Specifications

Antigen CALR
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 1G11-1A9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant CALR.
Formulation PBS with no preservative; pH 7.4
Gene CALR
Gene Accession No. BC020493
Gene Alias CRT/FLJ26680/RO/SSA/cC1qR
Gene Symbols CALR
Host Species Mouse
Immunogen CALR (AAH02500.1, 1 a.a. ∼ 417 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 811
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.