Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Calreticulin-2, Alexa Fluor 488, Clone: 321007, R&D Systems™

Catalog No. IC2927G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC2927G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC2927G Supplier R&D Systems Supplier No. IC2927G
This item is not returnable. View return policy

Applications:

Flow Cytometry (Intracellular Staining)

Specifications

Antigen Calreticulin-2/CALR3
Applications Flow Cytometry
Classification Monoclonal
Clone 321007
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q96L12
Gene Alias CALR3, calreticulin 2, calreticulin 3, Calreticulin2, Calreticulin-2, calreticulin-3, cancer/testis antigen 93, Crt2, CRT2calreticulin-2, CT93, FLJ25355, MGC26577
Gene Symbols CALR3
Host Species Mouse
Immunogen E. coli-derived recombinant human Calreticulin-2, Thr20-Leu384, Accession # Q96L12
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125972
Test Specificity Detects human Calreticulin-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Calreticulin is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.