Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calreticulin Mouse anti-Human, Clone: 16B11.1, MilliporeSigma™

Catalog No. MABT217MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT217MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT217MI Supplier MilliporeSigma Supplier No. MABT217
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Calreticulin Antibody, clone 16B11.1 is validated for use in Western Blotting, immunohistochemistry, Flow Cytometry for the detection of Calreticulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calreticulin
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 16B11.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_004334
Gene Symbols CALR, CRTC
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Calreticulin.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.