Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calreticulin Rabbit anti-Bovine, Human, Mouse, Pig, Rat, Xenopus, Polyclonal, MilliporeSigma™

Catalog No. 06661MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-661-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-661-MI Supplier MilliporeSigma Supplier No. 06661
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Calreticulin Antibody is an antibody against Calreticulin for use in immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calreticulin
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.0, 0.15M NaCl and 0.05% sodium azide.
Gene Symbols CALR; calreticulin; cC1qR; CRTC; HACBP; CRP55; grp60; FLJ26680; Calregulin; ERp60; RO; SSA
Host Species Rabbit
Immunogen peptide (QAKDEL) corresponding to the C-terminus (amino acids 412-417) of human calreticulin
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_004343
Target Species Bovine, Human, Mouse, Pig, Rat, Xenopus species
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.