Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calretinin Mouse anti-Mouse, Rat, Clone: 6B8.2, MilliporeSigma™

Catalog No. MAB1568MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1568MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1568MI Supplier MilliporeSigma Supplier No. MAB1568
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Calretinin Antibody, clone 6B8.2 detects level of Calretinin and has been published and validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calretinin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6B8.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing 0.02MPhosphate buffer, 0.25MNaCl, pH 7.6 with 0.1% sodium azide.
Gene Accession No. NM_001740.3
Gene Symbols CALB2, Calretinin, CAB29, CR, CAL2
Host Species Mouse
Immunogen Recombinant rat calretinin (Heinze, L., 2007).
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.