Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Calretinin, Clone: 6B8.2, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB1568/DEL

Description
Immunohistochemistry, Western Blotting

Specifications
Calretinin | |
Unconjugated | |
Recombinant rat calretinin | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
100 μg | |
Neuroscience | |
Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction