Learn More
CaM Kinase II Rabbit anti-Human, Rat, Mouse, Bovine, Hamster, Guinea Pig, Chicken, Rabbit, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | CaM Kinase II |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal IgG in buffer containing borate buffered saline and phosphate buffered saline containing 0.05% sodium azide. |
| Gene Symbols | CAMK2A; KIAA0968; CAMKA; EC 2.7.11.17 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to the N-terminus of Rat CaMK II |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.