Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase II Mouse, Unlabeled, Clone: 45, BD

Catalog No. BDB611292 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611292 50 μg
BDB611293 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611292 Supplier BD Biosciences Supplier No. 611292
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Ca2+;/calmodulin-dependent protein kinase II (CaM kinase II) is a multifunctional Ser/Thr kinase that regulates a number of cellular functions in response to increased intracellular Ca2+;. CaM kinase II is widely distributed, but is predominantly expressed in brain. It is involved in the regulation of neuronal functions such as neurotransmitter synthesis, neurotransmitter release, long-term potentiation, and formation of spatial learning. Neuronal CaM kinase II contains heteromers of two major subunits, α and β, at a ratio of 2:1 and homomers of α subunits. Each subunit has N-terminal ATP-binding and catalytic/regulatory domains and a C-terminal association domain. The regulatory domain consists of the autoinhibitory and calmodulin-binding sites. Assembly of the association domains of multiple subunits positions the regulatory domains for intersubunit autophosphorylation. After binding Ca2+;/calmodulin, CaM kinase II undergoes rapid autophosphorylation of the α and β subunits, which results in a substantial increase in its affinity for Ca2+;/calmodulin.

Host Species: Mouse
Clone: 45
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat CaM Kinase IIα aa. 448-460
Formula Weight [Chemical]: 52kDa

Immunofluorescence, Immunohistochemistry, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CaM Kinase II
Applications In vitro Assay, Western Blot
Classification Monoclonal
Clone 45
Concentration 250μg/mL
Conjugate Unconjugated
Description Ca2+/calmodulin-dependent protein kinase II
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat CaM Kinase IIα aa. 448-460
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.