Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase II Mouse anti-Bovine, Rat, Clone: 6G9, MilliporeSigma™

Catalog No. 05532MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-532-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-532-MI Supplier MilliporeSigma Supplier No. 05532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-CaM Kinase II Antibody, α subunit, clone 6G9 is validated for use in immunohistochemistry, western blotting for the detection of CaM Kinase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaM Kinase II
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6G9
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% azide before the addition of glycerol to 30%
Gene Accession No. NM_015981
Gene Symbols CAMK2A, KIAA0968, CAMKA
Host Species Mouse
Immunogen Partially purified rat CaM Kinase II, α-subunit
Purification Method Protein G chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.