Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase IV Mouse, Unlabeled, Clone: 26, BD

Catalog No. BDB610276 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610276 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610276 Supplier BD Biosciences Supplier No. 610276

Mouse Monoclonal Antibody

CaM-kinase IV (Ca2+calmodulin-dependent protein kinase IV, also described as CaM-kinase Gr) is activated through the binding of Ca2+/CaM and by phosphorylation. This kinase has high sequence homologies with the catalytic and regulatory domains of CaM-kinase II. CaM-kinase IV is a monomer which is highly expressed in cerebellum, thymus, and testis. Its subcellular distrubution includes localization in both the synaptic regions of the molecular layer of the cerebellum, and the nuclei of cerebellar granule cells. CaM-kinase IV has an autoinhibitory domain within residues 305-321 that can suppress kinase activity in the absence of Ca2+/CaM. This type of domain is common in many other CaM-dependent kinases and phosphatases. CaM-kinase IV does not appear to be significantly activated by autophosphorylation but it can be activated approximately 5-10-fold when phosphorylated by CaM-kinase IV kinase.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CaM Kinase IV
Applications Western Blot
Classification Monoclonal
Clone 26
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.