Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase Kinase Mouse, Unlabeled, Clone: 6, BD

Catalog No. BDB610544 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610544 50 μg
BDB610545 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610544 Supplier BD Biosciences Supplier No. 610544
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ca2+/Calmodulin (CaM)-dependent kinases are multifunctional kinases involved in a myriad of cellular functions such as neurotransmitter synthesis, long-term potentiation, and formation of spatial learning. A 68kDa CaM Kinase Kinase with homology to the Ca2+/CaM-dependent kinases, phosphorylates and activates Ca2+/CaM-dependent protein kinases I and IV, but not type II. This phosphorylation occurs on Ser/Thr residues and is Ca2+ and CaM dependent. CaM Kinase Kinase is abundant in brain tissue, and is thought to be an upstream regulator in the Ca2+/CaM-dependent neural processes.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CaM Kinase Kinase
Applications Western Blot
Classification Monoclonal
Clone 6
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.