Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

calcium/calmodulin-dependent protein kinase II delta, Mouse, Clone: 2E1, Abnova™

Catalog No. 89000555 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-555 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-555 Supplier Abnova Corporation Supplier No. H00000817M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CAMK2D.

The product of this gene belongs to the serine/threonine protein kinase family and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells, the enzyme is composed of four different chains: alpha, beta, gamma, and delta. The product of this gene is a delta chain. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Distinct isoforms of this chain have different expression patterns

Sequence: KGAILTTMLATRNFSAAKSLLKKPDGVKESTESSNTTIEDEDVKARKQEIIKVTEQLIEAINNGDFEAYTKICDPGLTAFEPEALGNLVEGMDFHRFYFENALSKSNKPI

Specifications

Antigen calcium/calmodulin-dependent protein kinase II delta
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2E1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CAMK2D.
Formulation PBS with no preservative; pH 7.4
Gene CAMK2D
Gene Accession No. BC032784
Gene Alias CAMKD/DKFZp686G23119/DKFZp686I2288/MGC44911
Gene Symbols CAMK2D
Host Species Mouse
Immunogen CAMK2D (AAH32784, 301 a.a. ∼ 410 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 817
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.