Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMK4 Mouse anti-Human, Clone: 8C5B8, MilliporeSigma™

Catalog No. MABF505MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF505MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF505MI Supplier MilliporeSigma Supplier No. MABF505
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-CAMK4, clone 8C5B8 Antibody is validated for use in western blotting, immunohistochemistry, immunofluorescence and ELISA for the detection of CAMK4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAMK4
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8C5B8
Conjugate Unconjugated
Formulation Purified mouse monoclonal in PBS containing up to 0.1% sodium azide and 0.5% protein stabilizer.
Gene Symbols CAMK4, CAMK, CAMK-GR, CAMKIV
Host Species Mouse
Immunogen Purified recombinant fragment of human CAMK4 expressed in E. Coli
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.