Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaMKI gamma Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA519661
Description
This antibody is predicted to react with rat and zebrafish based on sequence homology.
This gene encodes a protein similar to calcium/calmodulin dependent protein kinase, however, its exact function is not known.
Specifications
CaMKI gamma | |
Polyclonal | |
Unconjugated | |
Q91VB2, Q7TNJ7 | |
CAMK1G | |
Synthetic peptide conjugated to KLH derived from within residues 400 to the C-terminus of Mouse CaMKI gamma. | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.2 mg/mL | |
Camk1g | |
RP1-272L16.2, CLICKIII, VWS1, dJ272L16.1, CLICK III; caM kinase I gamma; caM kinase IG; caM-KI gamma; caMK-like CREB kinase III; caMKI gamma; caMKIG; calcium/calmodulin-dependent protein kinase type 1G | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
171358, 215303 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction