Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cAMP Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. 071497MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-149-7MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-149-7MI Supplier MilliporeSigma Supplier No. 071497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-cAMP Antibody detects level of cAMP and has been published and validated for use in immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen cAMP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Diluted serum presented as liquid in PBS containing no preservatives.
Host Species Rabbit
Immunogen 2'O-succinyl cyclic AMP conjugated to BSA
Purification Method Unpurified rabbit serum
Quantity 50 μL
Primary or Secondary Primary
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for long term. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.