Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
cullin-associated and neddylation-dissociated 1, Mouse, Clone: 5F2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CAND1.
Supplier: Abnova Corporation H00055832M02
Description
Sequence: MASASYHISNLLEKMTSSDKDFRFMATNDLMTELQKDSIKLDDDSERKVVKMILKLLEDKNGEVQNLAVKCLGPLVSKVKEYQVETIVDTLCTNMLSDKESpecifications
cullin-associated and neddylation-dissociated 1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_018448 | |
CAND1 | |
CAND1 (NP_060918, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Transcription Regulation | |
Primary | |
Human, Mouse, Rat | |
Antibody |
ELISA, Western Blot | |
5F2 | |
Mouse monoclonal antibody raised against a partial recombinant CAND1. | |
CAND1 | |
DKFZp434M1414/FLJ10114/FLJ10929/FLJ38691/FLJ90441/KIAA0829/TIP120/TIP120A | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
55832 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction