Learn More
Cannabinoid Receptor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Cannabinoid Receptor 1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity Purified immunoglobulin. Liquid in 100mM Tris, pH 7.5, containing 0.2% BSA and 0.05% sodium azide. |
| Gene Symbols | CNR1; CNR; CANN6; CB1A; CB1; CB1K5; CB-R |
| Host Species | Rabbit |
| Immunogen | A 14 amino acid N-terminus peptide sequence (aa 1-14) (Gerard et al. 1991; Gerard et al. 1990; Shire et al. 1995; Matsuda et al. 1990; Ho et al. 1996; Chakraborty et al. 1995). |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.