Learn More
Cannabinoid Receptor 1 Rabbit anti-Human, Monkey, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Cannabinoid Receptor 1 |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS, pH 7.7 with 0.01% sodium azide. |
| Gene Symbols | CNR1; CNR; CB1K5; CANN6; CB1A; CB-R; CB1 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide from the 3rd cytoplasmic domain of human Cannabinoid Receptor 1. The immunogen sequence has 100% homology with mouse. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.