Learn More
Cannabinoid Receptor 2 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Cannabinoid Receptor 2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in phosphate buffered saline, pH 7.4, containing 0.1% BSA and 0.1% sodium azide. |
| Gene Symbols | CNR2; CB-2; OTTHUMP00000044841; CB2; CX5 |
| Host Species | Rabbit |
| Immunogen | An 18 amino acid peptide sequence near the cytoplasmic, C-terminus of rat CB2 (Munro et al. 1993; Shire et al. 1996; Griffin et al. 2000). |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.