Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Cannabinoid Receptor CB1 (1-77), Polyclonal

Catalog No. 209550100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
20-955-0100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-955-0100UL Supplier MilliporeSigma™ Supplier No. 209550100UL

Rabbit Polyclonal Antibody

Specifically detects Cannabinoid Receptor CB1 (1-77) Clone: in Chicken, Fish, Gerbil, Human, Monkey, Mouse, Rat, Salamander samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Paraffin)

Cannabinoids exert their physiological effects through two G-protein coupled receptors, CB1 and CB2. Both receptors inhibit adenylyl cyclase and stimulate MAP kinase. CB1 receptors also modulate ion channels through direct G protein interactions. D-9-tetrahydrocannibinol and related ligands exert their psychoactive effects by inhibiting presynaptic N- and P/Q-type calcium channels. CB1 receptors are primarily expressed in brain (particularly cortex, striatum, hippocampus and cerebellum), spinal cord, eye, and testis. CB1 receptors are involved in the modulation of nociception, movement, learning and memory, emotion, and many other neuronal processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cannabinoid Receptor CB1 (1-77)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, 1mg/mL BSA, 50% glycerol.
Host Species Rabbit
Immunogen a recombinant protein consisting of the first 77 amino acids of rat CB1 receptor
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽60kDa CB1 receptor protein.
Target Species Chicken, Fish, Gerbil, Human, Monkey, Mouse, Rat, Salamander
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.