Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPN3, Mouse, Clone: 1B2, Abnova™

Catalog No. 89016425 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-425 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-425 Supplier Abnova Corporation Supplier No. H00000825M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CAPN3.

Calpain, a heterodimer consisting of a large and a small subunit, is a major intracellular protease, although its function has not been well established. This gene encodes a muscle-specific member of the calpain large subunit family that specifically binds to titin. Mutations in this gene are associated with limb-girdle muscular dystrophies type 2A. Alternate promoters and alternative splicing result in multiple transcript variants encoding different isoforms and some variants are ubiquitously expressed. [provided by RefSeq

Sequence: NKIKAWQKIFKHYDTDQSGTINSYEMRNAVNDAGFHLNNQLYDIITMRYADKHMNIDFDSFICCFVRLEGMFRAFHAFDKDGDGIIKLNVLEWLQLTMYA

Specifications

Antigen CAPN3
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CAPN3.
Formulation PBS with no preservative; pH 7.4
Gene CAPN3
Gene Accession No. BC003169
Gene Alias CANP3/CANPL3/LGMD2/LGMD2A/MGC10767/MGC11121/MGC14344/MGC4403/nCL-1/p94
Gene Symbols CAPN3
Host Species Mouse
Immunogen CAPN3 (AAH03169.1, 210 a.a. ∼ 309 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 825
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.