Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPN5, Mouse, Clone: 3C10, Abnova™

Catalog No. 89016383 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-383 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-383 Supplier Abnova Corporation Supplier No. H00000726M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CAPN5.

Calpains are calcium-dependent cysteine proteases involved in signal transduction in a variety of cellular processes. A functional calpain protein consists of an invariant small subunit and 1 of a family of large subunits. CAPN5 is one of the large subunits. Unlike some of the calpains, CAPN5 and CAPN6 lack a calmodulin-like domain IV. Because of the significant similarity to Caenorhabditis elegans sex determination gene tra-3, CAPN5 is also called as HTRA3. [provided by RefSeq

Sequence: VLGAAGLKDSPTGANSYVIIKCEGDKVRSAVQKGTSTPEYNVKGIFYRKKLSQPITVQVWNHRVLKDEFLGQVHLKADPDNLQALHTLHLRDRNSRQPSN

Specifications

Antigen CAPN5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CAPN5.
Formulation ascites with no preservative
Gene CAPN5
Gene Accession No. NM_004055
Gene Alias FLJ46245/HTRA3/nCL-3
Gene Symbols CAPN5
Host Species Mouse
Immunogen CAPN5 (NP_004046, 523 a.a. ∼ 622 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 726
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.