Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

calpain 5, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017460 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-460 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-460 Supplier Abnova Corporation Supplier No. H00000726D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human CAPN5 protein.

Calpains are calcium-dependent cysteine proteases involved in signal transduction in a variety of cellular processes. A functional calpain protein consists of an invariant small subunit and 1 of a family of large subunits. CAPN5 is one of the large subunits. Unlike some of the calpains, CAPN5 and CAPN6 lack a calmodulin-like domain IV. Because of the significant similarity to Caenorhabditis elegans sex determination gene tra-3, CAPN5 is also called as HTRA3. [provided by RefSeq

Sequence: MFSCVKPYEDQNYSALRRDCRRRKVLFEDPLFPATDDSLYYKGTPGPAVRWKRPKGICEDPRLFVDGISSHDLHQGQVGNCWFVAACSSLASRESLWQKVIPDWKEQEWDPEKPNAYAGIFHFHFWRFGEWVDVVIDDRLPTVNNQLIYCHSNSRNEFWCALVEKAYAKLAGCYQALDGGNTADALVDFTGGVSEPIDLTEGDFANDETKRNQLFERMLKVHSRGGLISASIKAVTAADMEARLACGLVKGHAYAVTDVRKVRLGHGLLAFFKSEKLDMIRLRNPWGEREWNGPWSDTSEEWQKVSKSEREKMGVTVQDDGEFWMTFEDVCRYFTDIIKCRVINTSHLSIHKTWEEARLHGAWTLHEDPRQNRGGGCINHKDTFFQNPQYIFEVKKPEDEVLICIQQRPKRSTRREGKGENLAIGFDIYKVEENRQYRMHSLQHKAASSIYINSRSVFLRTDQPEGRYVIIPTTFEPGHTGEFLLRVFTDVPSNCRELRLDEPPHTCWSSLCGYPQLVTQVHVLGAAGLKDSPTGANSYVIIKCEGDKVRSAVQKGTSTPEYNVKGIFYRKKLSQPITVQVWNHRVLKDEFLGQVHLKADPDNLQALHTLHLRDRNSRQPSNLPGTVAVHILSSTSLMAV

Specifications

Antigen calpain 5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human CAPN5 protein.
Formulation PBS with no preservative; pH 7.4
Gene CAPN5
Gene Accession No. NM_004055.4
Gene Alias FLJ46245/HTRA3/nCL-3
Gene Symbols CAPN5
Host Species Rabbit
Immunogen CAPN5 (NP_004046.2, 1 a.a. ∼ 640 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 726
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.