Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

calpain, small subunit 1, Mouse, Clone: 3C4, Abnova™

Catalog No. 89014066 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-066 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-066 Supplier Abnova Corporation Supplier No. H00000826M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CAPNS1.

Calpains are a ubiquitous, well-conserved family of calcium-dependent, cysteine proteases. Calpain families have been implicated in neurodegenerative processes, as their activation can be triggered by calcium influx and oxidative stress. Calpain I and II are heterodimeric with distinct large subunits associated with common small subunits, all of which are encoded by different genes. This gene encodes a small subunit common to both calpain I and II and is associated with myotonic dystrophy. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq

Sequence: KRWQAIYKQFDTDRSGTICSSELPGAFEAAGFHLNEHLYNMIIRRYSDESGNMDFDNFISCLVRLDAMFRAFKSLDKDGTGQIQVNIQE

Specifications

Antigen calpain, small subunit 1
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), KnockDown, Western Blot
Classification Monoclonal
Clone 3C4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CAPNS1.
Formulation PBS with no preservative; pH 7.4
Gene CAPNS1
Gene Accession No. BC000592
Gene Alias 30K/CALPAIN4/CANP/CANPS/CAPN4/CDPS
Gene Symbols CAPNS1
Host Species Mouse
Immunogen CAPNS1 (AAH00592, 172 a.a. ∼ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 826
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.