Learn More
CAPS, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human CAPS peptide using ARM Technology.
Supplier: Abnova Corporation H00000828K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a calcium-binding protein, which may play a role in the regulation of ion transport. A similar protein was first described as a potentially important regulatory protein in the dog thyroid and was termed as R2D5 antigen in rabbit. Alternative splicing of this gene generates two transcript variants. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
CAPS | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
CAPS | |
CAPS | |
A synthetic peptide of human CAPS | |
RUO | |
828 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
calcyphosine | |
CAPS | |
CAPS1, MGC126562 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.