Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capsaicin Receptor Guinea Pig anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5566MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5566MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5566MI Supplier MilliporeSigma Supplier No. AB5566
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Anti-Capsaicin Receptor Antibody, C-terminus detects level of Capsaicin Receptor and has been published and validated for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Capsaicin Receptor
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum. Liquid. Contains no preservative.
Gene Symbols TRPV1; VR1; OTRPC1; DKFZp434K0220; TrpV1
Host Species Guinea Pig
Immunogen A 22 amino acid peptide corresponding to the C-terminus of the rat Capsaicin Receptor protein (Catalog number AG298).
Purification Method Unpurified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_018727.5; NM_080704.3; NM_080705.3; NM_080706.3
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.