Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capsaicin Receptor Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB9554MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9554MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9554MI Supplier MilliporeSigma Supplier No. AB9554

Rabbit Polyclonal Antibody

Anti-Capsaicin Receptor Antibody is an antibody against Capsaicin Receptor for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Capsaicin Receptor
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Liquid.
Gene Symbols TRPV1; VR1; OTRPC1; TrpV1; DKFZp434K0220
Host Species Rabbit
Immunogen Synthetic peptide from the C-terminal of mouse VR1.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_018727.5; NM_080704.3; NM_080705.3; NM_080706.3
Target Species Mouse
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.