Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capsaicin Receptor Rabbit anti-Mouse, Rat, Primate, Polyclonal, MilliporeSigma™

Catalog No. AB5889MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5889MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5889MI Supplier MilliporeSigma Supplier No. AB5889
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Capsaicin Receptor Antibody, N-terminus is validated for use in western blot, immunohistochemistry for the detection of Capsaicin Receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Capsaicin Receptor
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid. Contains 0.05% sodium azide.
Gene Symbols TRPV1; VR1; OTRPC1; TrpV1; DKFZp434K0220
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminus of rat VR1.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_018727.5; NM_080704.3; NM_080705.3; NM_080706.3
Target Species Mouse, Primate, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.