Learn More
Capsaicin Receptor Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Capsaicin Receptor |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Serum. Lyophilized. Contains no preservative. Reconstitute with 50μL of sterile distilled water. Centrifuge if necessary to remove any residue. |
| Gene Symbols | TRPV1; VR1; OTRPC1; DKFZp434K0220; TrpV1 |
| Host Species | Rabbit |
| Immunogen | A 21 amino acid peptide corresponding to the C-terminus of the rat Capsaicin Receptor protein. |
| Quantity | 50 μL |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.