Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

capping protein (actin filament) muscle Z-line, alpha 1 (A02), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89004042 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-042 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-042 Supplier Abnova Corporation Supplier No. H00000829A02
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant CAPZA1.

CAPZA1 is a member of the F-actin capping protein alpha subunit family. This gene encodes the alpha subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments. [provided by RefSeq

Sequence: LGNSRFLDPRNKISFKFDHLRKEASDPQPEEADGGLKSWRESCDSALRAYVKDHYSNGFCTVYAKTIDGQQTIIACIESHQFQPKNFWN

Specifications

Antigen capping protein (actin filament) muscle Z-line, alpha 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant CAPZA1.
Formulation 50% glycerol
Gene CAPZA1
Gene Accession No. NM_006135
Gene Alias CAPPA1/CAPZ/CAZ1
Gene Symbols CAPZA1
Host Species Mouse
Immunogen CAPZA1 (NP_006126, 82 a.a. ∼ 170 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 829
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.