Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

capping protein (actin filament) muscle Z-line, alpha 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018800 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-800 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-800 Supplier Abnova Corporation Supplier No. H00093661D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human CAPZA3 protein.

This gene encodes an actin capping protein, one of the F-actin capping protein alpha subunit family. The encoded protein is predominantly localized to the neck region of ejaculated sperm, other immunohistochemical signals were found in the tail and postacrosomal regions. The encoded protein may also form heterodimers of alpha and beta subunits. This protein may be important in determining sperm architecture and male fertility. [provided by RefSeq

Sequence: MTLSVLSRKDKERVIRRLLLQAPPGEFVNAFDDLCLLIRDEKLMHHQGECAGHQHCQKYSVPLCIDGNPVLLSHHNVMGDYRFFDHQSKLSFKYYLLQNQLKDIQSHGIIQNEAEYLRVVLLCALKLYVNDHYPKGNCNMLRKTVKSKEYLIACIEDHNYETGECWNGLWKSKWIFQVNPFLTQVTGRIFVQAHFFRCVNLHIEISKDLKESLEIVNQAQLALSFARLVEEQENKFQAAVLEELQELSNEALRKILRRDLPVTRTLIDWHRILSDLNLVMYPKLGYVIYSRSVLCNWII

Specifications

Antigen capping protein (actin filament) muscle Z-line, alpha 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human CAPZA3 protein.
Formulation PBS with no preservative; pH 7.4
Gene CAPZA3
Gene Accession No. BC016745.2
Gene Alias CAPPA3/Gsg3
Gene Symbols CAPZA3
Host Species Rabbit
Immunogen CAPZA3 (AAH16745.1, 1 a.a. ∼ 299 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93661
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.