Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAR Mouse anti-Human, Mouse, Rat, Clone: RmcB, MilliporeSigma™

Catalog No. 05644MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-644-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-644-MI Supplier MilliporeSigma Supplier No. 05644
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CAR Antibody, clone RmcB detects level of CAR and has been published and validated for use in IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAR
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone RmcB
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing 0.1M Tris-Glycine, 0.15M NaCl and 0.05% Sodium Azide, pH 7.4.
Gene Accession No. NM_001338
Gene Symbols CXADR, CAR, hCAR, HCVADR, HCAR
Host Species Mouse
Immunogen Native human coxsackie virus-adenovirus receptor protein.
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.