Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-CAR, Clone: RmcB, ™ (Upstate™)
Supplier: MilliporeSigma™ 05644/DEL

Description
Immunocytochemistry, Western Blotting

Specifications
CAR | |
Unconjugated | |
Native human coxsackie virus-adenovirus receptor protein | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
200 μg | |
Cell Structure | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction