Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHST1 Monoclonal Antibody (543128)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524243
Description
In direct ELISAs, approximately 25-100% cross-reactivity with recombinant mouse CHST1 and no cross-reactivity with recombinant human CHST2, 3, 4, 5, 6, 7, 10, and 14 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Carbohydrate sulfotransferase 1 (CHST1), also referred to as KSGal6ST or KSST, is a keratan sulfate sulfotransferase. It is predominantly expressed in brain and skeletal muscle and localizes to the trans-Golgi network. CHST1 is responsible for mediating the sulfation of keratan in the cornea, which is important in maintaining corneal transparency. In particular, CHST1 catalyzes the transfer of sulfate groups from 3'-phosphoadenosine 5'-phosphosulfate to position six of internal or terminal galactose residues (preferentially the residues adjacent to sulfated GlcNAc) on growing keratan sulfate chains. CHST1 also contributes to the generation of L-selectin ligands. Mutations in the CHST1 gene may play a role in macular corneal dystrophy (MCD).
Specifications
| CHST1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O43916 | |
| CHST1 | |
| CHO-derived recombinant human Carbohydrate Sulfotransferase 1/CHST1 Arg24-Ser411. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Neutralization | |
| 543128 | |
| Unconjugated | |
| CHST1 | |
| 2610008E20Rik; AW125896; C6ST; carbohydrate (chondroitin 6/keratan) sulfotransferase 1; carbohydrate (keratan sulfate Gal-6) sulfotransferase 1; carbohydrate sulfotransferase 1; chst1; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 1; Gst1; GST-1; hypothetical protein LOC445124; keratan sulfate Gal-6 sulfotransferase; Keratan sulfotransferase; KS6ST; KSGal6ST; KSST; N-acetylglucosamine-6-O-sulfotransferase 1; sulfo1; sulfotransferase1; zgc:100904; zgc:100904 protein | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 8534 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction