Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase IV/CA4 Rat anti-Mouse, Clone: 565112, R&D Systems™

Catalog No. MAB2414
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2414 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2414 Supplier R&D Systems Supplier No. MAB2414
This item is not returnable. View return policy

Rat Monoclonal Antibody

Carbonic Anhydrase IV/CA4 Monoclonal antibody MAB2414 Specifically detects Carbonic Anhydrase IV/CA4 in Mouse samples. It is validated for Immunohistochemistry

Specifications

Antigen Carbonic Anhydrase IV/CA4
Applications Immunohistochemistry
Classification Monoclonal
Clone 565112
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q64444
Gene Alias CA4, CAIV, CA-IV, Car4, Carbonate dehydratase IV, carbonic anhydrase 4, carbonic anhydrase IVRP17, carbonic dehydratase IV, EC 4.2.1.1, retinitis pigmentosa 17 (autosomal dominant), RP17
Gene Symbols Car4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Carbonic Anhydrase IV, Glu18-Ser277, Accession # Q64444
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 762
Test Specificity Detects mouse CA4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CA1, 2, 3, 4, 5A, 5B, 6, 7, 8, 9, 10, 11, 12, 13, 14, recombinant mouse CA9, 12, or 14 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.