Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase H Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5314MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5314MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5314MI Supplier MilliporeSigma Supplier No. AB5314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Carboxypeptidase H Antibody, C-terminus is an antibody against Carboxypeptidase H for use in immunoprecipitation, western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxypeptidase H
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum with 0.02% sodium azide.
Gene Symbols CPE; CPH; EC 3.4.17.10
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminus of Carboxypepdidase H conjugated to KLH.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001873.1
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.