Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

caspase recruitment domain family, member 14, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008157 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-157 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-157 Supplier Abnova Corporation Supplier No. H00079092B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human CARD14 protein.

The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein shares a similar domain structure with CARD11 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10. Two alternatively spliced variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq

Sequence: MVSSCELELQEQSLRTASDQESGDEELNRLKEENEKLRSLTFSLAEKDILEQSLDEARGSRQELVERIHSLRERAVAAERQREQYWEEKEQTLLQFQKSKMACQLYREKVNALQAQVCELQKERDQAYSARDSAQREISQSLVEKDSLRRQVFELTDQVCELRTQLRQLQAEPPGVLKQEARTREPCPREKQRLVRMHAICPRDDSDCSLVSSTESQLLSDLSATSSRELVDSFRSSSPAPPSQQSLYKRVAEDFGEEPWSFSSCLEIPEGDPGALPGAKAGDPHLDYELLDTADLPQLESSLQPVSPGRLDVSESGVLMRRRPARRILSQVTMLAFQGDALLEQISVIGGNLTGIFIHRVTPGSAADQMALRPGTQIVMVSRARPLLSPGLLMGTVAAGGVTQADFTSPRRCRSTLGWASALSWADVKRSAHL

Specifications

Antigen caspase recruitment domain family, member 14
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human CARD14 protein.
Formulation No additive
Gene CARD14
Gene Accession No. NM_052819
Gene Alias BIMP2/CARMA2
Gene Symbols CARD14
Host Species Mouse
Immunogen CARD14 (NP_438170, 1 a.a. ∼ 434 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Apoptosis
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 79092
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.