Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CARP Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA3852
Description
Carbonic anhydrase-related protein (CARP) is a novel IP3R1-binding protein. Analysis of this protein revealed it is expressed exclusively in the Purkinje cells of the cerebellum.
PA3-852 detects CARP from human, mouse, and rat samples.PA3-852 has been successfully used in Western blot and immunocytochemistry procedures.The PA3-852 immunogen is a synthetic peptide corresponding to residues C(267) D G I L G D N F R P T Q(269) of mouse CARP.
Specifications
CARP | |
Polyclonal | |
1:1,000 | |
Q15327, Q9CR42, Q8R560 | |
ANKRD1 | |
Synthetic peptide corresponding to residues C(267) D G I L G D N F R P T Q(279) of mouse CARP. | |
RUO | |
107765, 27063, 27064 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
Unconjugated | |
ANKRD1 | |
Carbonic anhydrase-related protein | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction