Learn More
cancer susceptibility candidate 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CASC3.
Supplier: Abnova Corporation H00022794A01
Description
The product of this gene is a core component of the exon junction complex (EJC), a protein complex that is deposited on spliced mRNAs at exon-exon junctions and functions in nonsense-mediated mRNA decay (NMD). The encoded protein binds RNA and interacts with two other EJC core components. It is predominantly located in the cytoplasm, but shuttles into the nucleus where it localizes to nuclear speckles. [provided by RefSeq
Sequence: VSMSPGQPPPQQLLAPTYFSAPGVMNFGNPSYPYAPGALPPPPPPHLYPNTQAPSQVYGGVTYYNPAQQQVQPKPSPPRRTPQPVTIKPPPPEVVSRGSSSpecifications
cancer susceptibility candidate 3 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant CASC3. | |
CASC3 | |
BTZ/MLN51 | |
Mouse | |
50 μL | |
Yes | |
22794 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_007359 | |
CASC3 | |
CASC3 (NP_031385, 604 a.a. ∼ 703 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.