Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 2α Mouse anti-Human, Mouse, Rat, Clone: 1AD9, MilliporeSigma™

Catalog No. 051431MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-143-1MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-143-1MI Supplier MilliporeSigma Supplier No. 051431
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Casein Kinase 2α Antibody, 1AD9 detects level of Casein Kinase 2α and has been published and validated for use in immunocytochemistry, western blot, immunohistochemistry (paraffin) and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Casein Kinase 2α
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1AD9
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing 0.02MPhosphate, pH 7.6 with 0.25MNaCl and 0.1% sodium azide.
Gene Accession No. NM_001895.3
Gene Symbols CSNK2A1, CK2A1, CKII, CK II 3 EC 2.7.11.1
Host Species Mouse
Immunogen Purified recombinant α-subunit of casein kinase 2.
Purification Method Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.