Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Casein Kinase II, alpha-Subunit Clone: 1AD9,

Catalog No. 218703100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
21-870-3100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-870-3100UG Supplier MilliporeSigma™ Supplier No. 218703100UG

Mouse Monoclonal Antibody

Specifically detects Casein Kinase II, α-Subunit Clone: 1AD9 in Bovine, Hamster, Human, Mouse samples, and it is validated for Immunocytochemistry

Human Casein Kinase II (CK2, CK-II) is a serine-threonine-specific protein kinase comprised of two ∽45kDa catalytic (α) subunits and two ∽25kDa regulatory (β) subunits in an α2β2 tetramer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Casein Kinase II, α-Subunit
Applications Immunocytochemistry
Classification Monoclonal
Clone 1AD9
Conjugate Unconjugated
Formulation Lyophilized from PBS.
Host Species Mouse
Immunogen purified, recombinant, SDS-denatured, human CK2 α-subunit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽45kDa CK2α subunit in F9 cells.
Target Species Bovine, Hamster, Human, Mouse
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.