Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase IIα/α Mouse, Unlabeled, Clone: 31, BD

Catalog No. BDB611610 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611610 50 μg
BDB611611 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611610 Supplier BD Biosciences Supplier No. 611610

Mouse Monoclonal Antibody

Casein kinase II (CKII, CK2) is a pleiotropic, ubiquitous, and constitutively active Ser/Thr kinase that utilizes both ATP and GTP as phosphoryl donors to catalyze the phosphorylation of numerous protein substrates. It has been implicated in DNA replication, control of metabolism, and regulation of transcription and translation. CKII is a messenger independent kinase due to the fact that its activity is not affected by factors that regulate other protein kinases (e.g. cAMP, Ca2+, diacylglycerol). It is composed of two catalytic (α and α′) subunits and two regulatory (β) subunits. The β subunits stabilize the holoenzyme and enhance the activity of the α subunits. Although the enzyme exists as a tetramer, the free α/α′ subunits are catalytically active by themselves and are present in cells under some conditions. The α and α′ subunits (85% homologous) are encoded by distinct genes and have been detected in multiple species including human, bovine, and avian. CKII is found primarily in the nucleus in dividing cells, but can be detected in the cytoplasm in quiescent cells. Thus, CKII activity is required for cell cycle progression and other essential signal transduction events.

Host Species: Mouse
Clone: 31
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Rat Casein Kinase II α aa. 1-123

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Casein Kinase IIα/α
Applications Western Blot
Classification Monoclonal
Clone 31
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Casein Kinase II α aa. 1-123
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.