Learn More
Description
Casein kinase II (CKII, CK2) is a pleiotropic, ubiquitous, and constitutively active Ser/Thr kinase that utilizes both ATP and GTP as phosphoryl donors to catalyze the phosphorylation of numerous protein substrates. It has been implicated in DNA replication, control of metabolism, and regulation of transcription and translation. CKII is a messenger independent kinase due to the fact that its activity is not affected by factors that regulate other protein kinases (e.g. cAMP, Ca2+, diacylglycerol). It is composed of two catalytic (α and α′) subunits and two regulatory (β) subunits. The β subunits stabilize the holoenzyme and enhance the activity of the α subunits. Although the enzyme exists as a tetramer, the free α/α′ subunits are catalytically active by themselves and are present in cells under some conditions. The α and α′ subunits (85% homologous) are encoded by distinct genes and have been detected in multiple species including human, bovine, and avian. CKII is found primarily in the nucleus in dividing cells, but can be detected in the cytoplasm in quiescent cells. Thus, CKII activity is required for cell cycle progression and other essential signal transduction events.
Host Species: Mouse
Clone: 31
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Rat Casein Kinase II α aa. 1-123
Immunofluorescence, Western Blotting
Specifications
Specifications
| Antigen | Casein Kinase IIα/α |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 31 |
| Concentration | 250μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Rat Casein Kinase II α aa. 1-123 |
| Purification Method | Affinity Purified |
| Quantity | 150 μg |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.