Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASP2, Mouse, Clone: 1C11, Abnova™

Catalog No. 89016430 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-430 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-430 Supplier Abnova Corporation Supplier No. H00000835M10
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CASP2.

This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The proteolytic cleavage of this protein is induced by a variety of apoptotic stimuli. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms. [provided by RefSeq

Sequence: TLSGLQHVLPPLSCDYDLSLPFPVCESCPLYKKLRLSTDTVEHSLDNKDGPLCLQVKPCTPEFYQTHFQLAYRLQSRPRGLALVLSNVHFTGEKELEFRS

Specifications

Antigen CASP2
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1C11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CASP2.
Formulation PBS with no preservative; pH 7.4
Gene CASP2
Gene Accession No. BC002427
Gene Alias CASP-2/ICH-1L/ICH-1L/1S/ICH1/NEDD2
Gene Symbols CASP2
Host Species Mouse
Immunogen CASP2 (AAH02427, 121 a.a. ∼ 220 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 835
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.