Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caspase 12 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB3613
Description
This Anti-Caspase 12 Antibody, N-terminus is validated for use in western blotting for the detection of Caspase 12.
Specifications
Caspase 12 | |
Polyclonal | |
Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide. | |
Rabbit | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
CASP12; Caspase-12.; CASP12P1 | |
Peptide corresponding to aa 2-17 of mouse caspase-12. | |
Primary | |
4°C for 6 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction